Learn More
Liebherr™ Forced-Air Refrigerator with Alarm
Product Code. 12527116 Shop All Liebherr Products
Click to view available options
Capacity (Metric):
434 L
583 L
663 L
Temperature Range:
-2°C to +15°C
1°C to 15°C
Color:
Stainless Steel
White
Dimensions (L x W x H) Exterior:
1864 H x 750 W x 750 D mm
1900 H x 600 W x 680 D mm
2064 H x 750 W x 750 D mm
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 12527116
Brand: Liebherr™ GKv 4310
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Regulatory: This product is under eco-contribution tax.
Use these refrigerators for general laboratory storage of temperature sensitive samples. Liebherr™ Forced-Air Refrigerators feature plastic coated grid shelves and plastic interior containers.
  • Choice of white or stainless steel exterior
  • SwingLine door design with interchangeable hinges
  • Ergonomic slimline handle
  • Automatic defrost
  • Dynamic cooling system
  • Electronic control system
  • External digital temperature display
Specifications
Capacity (Metric) 434 L
Temperature Range 1°C to 15°C
Refrigerant R600A
Door Style Self-closing
Door Material Steel Door/Lid
Shelves 6
Cabinet Material Steel
Interior Plastic Interior Containers
Color White
Warranty 2 Years
Defrost Automatic
Model GKV4310 X
Dimensions (L x W x H) Exterior 1900 H x 600 W x 680 D mm
Depth (Metric) Interior 480 mm
Height (Metric) Interior 1688 mm
Width (Metric) Interior 475 mm
Voltage 220/240 V
Frequency 50 Hz
Shipping Weight (Metric) 73 kg