missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Forma™ -20°C Enzyme Freezer

Maintain consistent temperatures with these manual defrost enzyme freezers, which feature gravity convection air systems without blowers or fans.

POA

Specifications

Type Enzyme Freezer
Temperature Range -20°C
Door Style Solid
No. of Doors 1
Cabinet Material Scratch-resistant Painted
View More Specs

Description

Description

Maintain consistent temperatures with Thermo Scientific™ Forma™ -20°C Enzyme Freezers, which feature manual defrost and microprocessor control. Our Forma Enzyme Freezer is designed for storage applications where slight intermittent coil warming during an auto-defrost cycle is not desired. The gravity convection air system provides maximum stability without blowers or fans.

Microprocessor Control

  • Enhanced digital electronic technology simplifies operation and delivers greater accuracy
  • Operating temperature preset at -20°C
  • Icon-based controls and displays include graphic references
  • Setpoint security with key-operated triple-position master switch controls power to all systems
  • Audible/visual warnings for overtemperature, undertemperature and power failure
  • Visual status reports on critical functions
  • Graphic thermometer confirms operation: steady green for normal, flashing top bar for overtemperature, flashing bulb for undertemperature
  • Enlarged digital display with temperature resolution to within 0.1°C
  • Alarm silence, ringback and automatic reset functions
  • Pushbutton alarm test verifies sensor continuity
  • Battery backup
  • Remote alarm contacts

Refrigeration System

  • Heavy-duty, industrial-grade, hermetically sealed compressors
  • Manual defrost for maximum temperature uniformity performance

Construction

  • Industrial-quality cabinet construction
  • High-density, CFC-free urethane foam insulation
  • Solid door with keyed door lock
  • Door self-closes against closed-cell peripheral gasket
  • Full-length door handle
  • Adjustable leveling feet standard (optional casters available)
  • Scratch-resistant painted interior and exterior
  • Nine epoxy-coated, open-wire shelves, adjustable in 1 in. (2.5cm) increments—no tools required

Includes:

  • 45 enzyme storage bins (5 per shelf); labels not provided
  • Warranty varies by country, please contact us for details
Specifications

Specifications

Enzyme Freezer
Solid
Scratch-resistant Painted
Epoxy Coated Steel
Microprocessor
Manual
79.2 in.
37.2 in.
Alarm
CFC-free
-20°C
1
Urethane Foam
9
Warranty varies by country, please contact us for details
29 in.
58 in.
94.5 cm
Heavy-Duty, Industrial-Grade, Hermetically Sealed
Videos
SDS
Documents

Documents

Product Certifications