Learn More
Thermo Scientific™ Forma™ 88000 Series -86°C Upright Ultra-Low Temperature Freezers
Store the most critical samples in these upright ultra-low temperature freezers, which feature outstanding safety, security and thermal performance.
Description
Store the most critical samples in Thermo Scientific™Forma™ 88000 Series -86°C Upright Ultra-Low Temperature Freezers, which feature outstanding safety, security and thermal performance.
Features:- Choice of five capacities maximizes sample storage while minimizing the freezer's footprint; features the industry's leading capacity per footprint specifications‡
- High-performance mode provides the tightest temperature uniformity and peak variation†—for most applications, energy-savings mode offers excellent temperature control, plus up to 15% savings on energy usage compared to high performance mode
- Increases the internal capacity of 2 in. vials over previous generation freezers—gain up to 76% more capacity in the same footprint with 1mL CryoBank tubes†
- Monitor the freezer's health 24/7 and access a detailed event log—simply touch the heart icon on the main screen to see freezer's temperature, and access settings and operating parameters
- Download a report of the current event log to a portable drive
- Experience more efficient heat transfer with a brazed plate heat exchanger and joints to reduce leak potential and improve reliability
- Protect against a wide range of voltage variation with a power management system that is easily accessible through the touch-screen display
- Monitor the freezer with remote alarm contacts: 4-20 milliamp output compatible with external alarm and monitoring systems
- Use inexpedient probes or instrumentation with two 1 in. (25mm) rear access ports
- Store up to 15 years worth of temperature and event data on the on-board compute
- Use the USB port to download freezer temperature and event log data, or freezer settings from one freezer to another
- Protect against dust on the condenser, which can cause reduced refrigeration performance and increased risk to samples, with easy-to-remove, washable filter
- Store up to 278 lbs. of samples (depending on freezer model) on reinforced, stainless steel shelving
- Maximize cabinet temperature and eliminate frost build-up with 4×7 heated gasket with four touchpoints of security and seven zones of protection
- Maintain cabinet temperature during openings with five polystyrene insulated inner doors that feature embedded rare earth magnets, eliminating exposed latches or magnets
- Several optional features including LN2 and CO2 back-up system, chart recorder, stainless steel inventory racking solutions and hands-free locking option
- Environmentally-friendly, CFC/HCFC free refrigerants
- Warranty varies by country, please contact us for details
‡Based on internal performance data; Data on file May 2011 †Comparison of energy-savings and high-performance mode.