missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde solution, Honeywell™

Catalog Number p-7145545
Click to view available options
Quantity:
500 mL
1 L
2.5 L
This item is not returnable. View return policy
50-00-0, 67-56-1, 7732-18-5
CH2O
30.03
MFCD00003274
WSFSSNUMVMOOMR-UHFFFAOYSA-N
712
CHEBI:16842
formaldehyde
C=O
This item is not returnable. View return policy
50-00-0, 67-56-1, 7732-18-5
CH2O
30.03
MFCD00003274
WSFSSNUMVMOOMR-UHFFFAOYSA-N
712
CHEBI:16842
formaldehyde
C=O

ACS reagent, 37 wt.% in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY
CAS Min % ≥30.00%, ≥10.00%, ≥50.00%
CAS Max % <50.00%, <20.00%, <70.00%
Quantity 500 mL
Physical Form Liquid
Grade ACS general use solvents
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'