missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, Technical, Solution, Stabilised In Methanol, Fisher Chemical™

Catalog Number p-7073026
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
50-00-0
CH2O
30.03
MFCD00003274
WSFSSNUMVMOOMR-UHFFFAOYSA-N
712
CHEBI:16842
formaldehyde
C=O
50-00-0
CH2O
30.03
MFCD00003274
WSFSSNUMVMOOMR-UHFFFAOYSA-N
712
CHEBI:16842
formaldehyde
C=O
Physical Form Liquid
Formula Weight 30.02g/mol
Packaging HDPE plastic bottle
Vapor Pressure 2mbar at 20°C
Vapor Density > 1.0
pH 3 to 4.2
Melting Point -15°C
Boiling Point 97°C
Color Colorless
Quantity 2.5 L
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formaldehyde solution, 37%
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H370-Causes damage to organs.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Formaldehyde, Methylalcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'