Learn More
Poulten Graf™ Fortuna™ Glass 1 Mark Bulb Pipette
Click to view available options
Volume (Metric):
2 mL
10 mL
Unit Size:
Each
Pack of 5
Description
- Short practical delivery time
- Tips and suction tubes reinforced
- Acid and alkali resistant
Specifications
Specifications
| Class | AS |
| Material | Glass |
| Graduated | Yes |
| Graduation Color | Amber |
| Quantity | 1 Pack |
| Volume (Metric) | 10 mL |
| Type | Bulb Pipet |