Learn More
Poulten Graf™ Fortuna™ Optima™ All Glass Luer-Tip Syringe
Excellent for pharmaceutical, cosmetics and biotechnological industries. Poulten Graf™ Fortuna™ Optima™ All Glass Luer-Tip Syringes come with an interchangeable piston.
6.90€ - 9.65€
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | CE EN 46001 |
Material | Borosilicate Glass |
Description | Amber graduation |
Product Type | All Glass Luer-tip Syringe |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10281771
|
Poulten Graf™
7 10237 |
10 mL |
9.65€
Each |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10405461
|
Poulten Graf™
7 10227 |
2 mL |
6.90€
Each |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10333661
|
Poulten Graf™
7 10233 |
5 mL |
7.60€
Each |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Complies with CE EN 46001 standards
- Autoclavable at 134°C
- Constructed from Durobax™ glass
- With permanent amber graduation
- Acid and alkali resistant
Specifications
Autoclavable | |
Borosilicate Glass | |
All Glass Luer-tip Syringe | |
Luer Tip |
CE EN 46001 | |
Amber graduation | |
1/Ea. | |
Yes |