Learn More
Poulten & Graf GmbH FORTUNA™ Volumetric Flasks with PP-Stopper and Amber Graduation
Poulten & Graf GmbH FORTUNA™ Volumetric Flasks with PP-Stopper and Amber Graduation allows precise measurement of set volumes.
57.85€
Specifications
Certifications/Compliance | EN ISO 1042 |
---|---|
Closure Color | White |
Closure Material | Polypropylene |
Description | Volumetric flask with PP-stopper |
Material | Borosilicate Glass |
Description
- Amber graduations
- Conformity approved
- With tolerance indications
- Amber graduations are acid and alkali resistant
- With PP-Stopper
Specifications
EN ISO 1042 | |
Polypropylene | |
Borosilicate Glass | |
Volumetric flask with PP-stopper | |
Non-sterile | |
Flat |
White | |
Volumetric flask with PP-stopper | |
Amber Ring Marks | |
Reusable | |
A | |
Stopper |