missing translation for 'onlineSavingsMsg'
Learn More

Ohaus™ Four Block Analog Dry Block Heater HB4AL

Product Code. 15792699 Shop All Ohaus Products
Click to view available options
No. of Blocks:
4
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15792699

Brand: Ohaus™ 30392068

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

OHAUS™ Four Block Analog Dry Block Heater processes more samples while delivering precise and reliable results.

  • High wattage, constant-temperature Dry Block Heaters are economical, versatile and compact
  • These multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points and a wide variety of other laboratory procedures
  • Each of the five models accept separate interchangeable modular blocks (available separately), accommodating various sample enclosures such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes
  • Each block has a thermometer well for measuring block temperature
  • Anodised aluminum modular blocks provide superior temperature stability and heat transfer
  • Analogue controls
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance TUV
Controller Type Analog
Description Dry Block Heater, 4 Block, HB4AL, EU
Temperature Range (Metric) 5°C to 100°C (Low), 75°C to 150°C (High)
Type Dry Bath Block
Heat Up Time 70 min
Model HB4AL
No. of Blocks 4
Temperature Stability ±2°C
Temperature Uniformity ±0.1°C at 37°C
Uniformity ± 0.4°C at 37°C
Dimensions (L x W x H) 42.9 x 20.3 x 8.9 cm
Length (English) 16.90 in.
Length (Metric) 42.90 cm
Width (English) 8.00 in.
Width (Metric) 20.30 cm
Height (English) 3.50 in.
Height (Metric) 8.900 cm
Depth (English) 16.90 in.
Depth (Metric) 42.90 cm
Weight (Metric) 3.8 kg
Electrical Requirements 230V, 1.35A, 50/60Hz
Amperage 1.35A
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.