Learn More
Ohaus™ Four Block Digital Dry Block Heater HB4DG
Product Code. 15722709 Shop All Ohaus Products
Click to view available options
Controller Type:
Digital
Unit Size:
Each
Product Code. 15722709
Brand: Ohaus™ 30392089
Only null left
Add to basket
Add to basket
OHAUS™ Four Block Digital Dry Block Heater processes more samples while delivering precise and reliable results.
  • Designed for applications that require repeatable results and superior temperature stability
  • These multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a wide variety of other laboratory procedures
  • Integral support rod holder with locking knob accepts optional external temperature probe kit
  • Optional external temperature probe kit monitors actual block or sample temperature
  • Each of the five models accepts separate interchangeable modular blocks (available separately) accommodating various tube sizes from 0.2 mL microtubes to 50 mL centrifuge tubes
  • Each block has a thermometer well for measuring block temperature
  • Efficient heating due to close tube-and-block contact for maximum heat retention
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance TUV
Controller Type Digital
Description Dry Block Heater, 4 Block, HB4DG, EU
Temperature Range (Metric) 5°C to 120°C
Type Dry Bath Block
Heat Up Time 60 min
Model HB4DG
No. of Blocks 4
Temperature Stability ±0.1°C
Temperature Uniformity ±0.1°C at 37°C
Uniformity ± 0.2°C at 37°C
Dimensions (L x W) 42.90 cm x 20.30 cm
Dimensions (L x W x H) 42.9 x 20.3 x 8.9 cm
Length (English) 16.90 in.
Length (Metric) 42.90 cm
Width (English) 8.00 in.
Width (Metric) 20.30 cm
Height (English) 3.50 in.
Height (Metric) 8.900 cm
Depth (English) 16.90 in.
Depth (Metric) 42.90 cm
Weight (Metric) 3.8 kg
Electrical Requirements 230V, 1.35A, 50/60Hz
Amperage 1.35A