Learn More
Novus Biologicals™ Foxp3/ Transcription Factor Staining Buffer Set
Product Code. 18788033 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18788033 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18788033 Supplier Novus Biologicals™ Supplier No. NBP2266911
The Foxp3/Transcription Factor Staining Buffer Set have been formulated and optimized for staining with antibodies to transcription factors and nuclear proteins, such as Foxp3 and Ki-67 as well as cytokines and chemokines.
The Foxp3/Transcription Factor Staining Buffer Set have been formulated and optimized for staining with antibodies to transcription factors and nuclear proteins, such as Foxp3 and Ki-67 as well as cytokines and chemokines. Buffer: Solubilize the peptides prior to use by making 5 mM PBS stock solutions (please see Preparation of 5 mM Stock Solutions under Preparation Method).
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Intracellular staining followed by flow cytometric analysis
Quantity 1 Kit