Learn More
Hanna Instruments™ Free Chlorine Standard, 0.50mg/L
Quickly check the calibration of Hanna Free Chlorine Meter HI 93701 and HI 93711. Hanna Instruments™ Free Chlorine Standard, 0.50mg/L includes four (4) sealed cuvets of free chlorine standard, comprising 0.5 ppm Free Chlorine.
Brand:  Hanna Instruments™ HI9370150
Additional Details : Weight : 0.80000kg
Product Code. 11753554
  • 312.00€ / Pack of 4
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
Specifications
Free chlorine determination
4 sealed cuvets of free chlorine standard
4 Bottles
Calibration standard
HI 93701 and HI 93711 Free Chlorine Meters
4
0.50mg/L (ppm)