Learn More
Hanna Instruments™ Free and Total Chlorine Portable Photometer
Click to view available options
Range:
0 to 5mg/L
Length (Metric):
192 mm
193 mm
Unit Size:
Each
Specifications
Specifications
| Type | Portable Photometer |
| Accuracy | ±0.03mg/L |
| For Use With (Application) | Free and Total Chlorine |
| Height (Metric) | 69 mm |
| Length (Metric) | 192 mm |
| Light Source | Tungsten Lamp |
| Range | 0 to 5mg/L |
| Resolution | 0.01mg/L |
| Weight (Metric) | 0.36 kg |
| Width (Metric) | 104 mm |