Learn More
Brady™ FREEZERBONDZ™ Polyester Labels
Product Code. 15159452
Click to view available options
Size (Metric):
12.7 mm dia.
48.3 W x 25.4 mm H
Unit Size:
Each
Product Code. 15159452
Brand: Brady™ 142894
For use in laboratory identification applications such as vials, centrifuge tubes, test tubes, straws and slides.
  • Polyester
  • Applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen
  • Provides excellent print smudge resistance, solvent resistance and low-temperature performance
Specifications
Certifications/Compliance RoHS compliant to 2002/618/EC MCV amendment to RoHS Directive 2011/65/EU
Description FreezerBondz Labels for the BBP33 Printer
Adhesive Type Acrylic, Permanent
Color White
Finish Matte
Format Die-cut
Material Polyester
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) For laboratory identification
For Use With (Equipment) BBP™ 33
Labels per Row 1
Min. Operating Temperature (Metric) -196°C
No. per Pack 1500/Pk.
Quantity 1500/Pk.
Shape Rectangle
Size (Metric) 48.3 W x 25.4 mm H
Software Type LabelMark, MarkWare
Temperature Range (English) -320°F to +266°F
Temperature Range (Metric) -196°C to +130°C
Thickness (English) 0.0026 in.
Thickness (Metric) 0.013 mm
Dimensions (LxW) 25.40 x 48.26 mm
Length (English) 1 in.
Width (English) 1.9 in.
Width (Metric) 48.26 mm
Height (English) 1 in.
Height (Metric) 25.4 mm
Alternate Part Number B33-156-490