missing translation for 'onlineSavingsMsg'
Learn More

Fruit Juice Organic Acid Mixture 56 25-2000 μg/mL in Water, Dr. Ehrenstorfer

Product Code. 30312953
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
This item is not returnable. View return policy

Product Code. 30312953

Brand: Dr. Ehrenstorfer DREGS09000056WA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Citric Acid∣D-(+)-Malic Acid∣DL-Tartaric acid∣Fumaric acid∣Quinic Acid
CAS 77-92-9, 636-61-3, 133-37-9, 110-17-8, 77-95-2
Analyte or Component Names Citric Acid∣D-(+)-Malic Acid∣DL-Tartaric acid∣Fumaric acid∣Quinic Acid
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Water
Quantity 5 x 1 mL
Concentration or Composition (by Analyte or Components) Fumaric acid 25 μg/mL∣Citric Acid 2000 μg/mL∣D-(+)-Malic Acid 2000 μg/mL∣DL-Tartaric acid 2000 μg/mL∣Quinic Acid 2000 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.