missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ FTBMT

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
FTBMT (also known as TP 024) is a potent and selective GPR52 agonist (EC50 = 75 nM, Emax = 122%), which is selective for GPR52 over a panel of 98 targets including D1, D2, AMPA and NMDA. FTBMT suppresses methamphetamine-induced hyperlocomotion in mice, and inhibits MK-801 -induced hyperactivity (model for acute psychosis), without causing catalepsy in mice. This compound is orally bioavailable and brain penetrant.
Specifications
Specifications
| Chemical Name or Material | 4-[3-[[3-Fluoro-5-(trifluoromethyl)phenyl]methyl]-5-methyl-1H-1,2,4-triazol-1-yl]-2-methylbenzamide |
| CAS | 1358575-02-6 |
| Quantity | 5 mg |
| Molecular Formula | C19H16F4N4O |
| Formula Weight | 392.35 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction