missing translation for 'onlineSavingsMsg'
Learn More

Full Pharma and Food Flex Kit for Metals Analysis, VHG™

Product Code. 30003095
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1200mL
Product Code. Quantity unitSize
30003095 1 Kit 1200mL
1 options
This item is not returnable. View return policy

Product Code. 30003095

Brand: VHG Labs VHGPHARMKIT12X100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Full Pharma and Food Flex Kit for Metals Analysis
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium/Antimony/Arsenic/Barium/Bismuth/Boron/Cadmium/Calcium/Chromium/Cobalt/Copper/Germanium/Gold/Indium/Iridium/Iron/Lead/Lithium/Lutetium/Magnesium/Manganese/Mercury/Molybdenum/Nickel/Osmium/Palladium/Platinum/Potassium/Rhodium/Ruthenium/Scandium/Selenium/Silver/Sodium/Tellurium/Thallium/Tin/Vanadium/Zinc
CAS 7429-90-5, 7440-36-0, 7440-38-2, 7440-39-3, 7440-69-9, 7440-42-8
Methods ICP/ICP-MS
Analyte or Component Names Aluminium/Antimony/Arsenic/Barium/Bismuth/Boron/Cadmium/Calcium/Chromium/Cobalt/Copper/Germanium/Gold/Indium/Iridium/Iron/Lead/Lithium/Lutetium/Magnesium/Manganese/Mercury/Molybdenum/Nickel/Osmium/Palladium/Platinum/Potassium/Rhodium/Ruthenium/Scandium/Selenium/Silver/Sodium/Tellurium/Thallium/Tin/Vanadium/Zinc
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.