Learn More
Saint Gobain Life Sciences Chemware™ PFA Funnel
Click to view available options
Diameter (Metric)Top:
95 mm
Unit Size:
Each
Description
- Non-contaminating
- Inert
- Non-stick surface reduces residual material left in funnel
- The top cone has a high side to limit splashing, while the narrow step fits into a range of containers
- The low surface energy of PFA facilitates fast, complete drainage and easy clean up
Specifications
Specifications
| Diameter (Metric)Top | 95 mm |
| Material | PFA |
| Height (Metric) | 112 mm |
| Diameter (English)Top | 3.7 in. |
| Height (English) | 4.4 in. |
| Product Type | Funnel |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion