Learn More
Pyrex™ Funnel with Porous Plate
Click to view available options
Capacity (Metric):
15 mL
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric)Top | 3mm |
| Capacity (Metric) | 15 mL |
| For Use With (Application) | HPLC |
| Pore Size | 130Å |
| Product Type | Funnel |