Learn More
Usbeck Laborgeraete™ Funnels
Click to view available options
Height (Metric):
120 mm
170 mm
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric)Top | 150mm |
| Material | Stainless Steel |
| Height (Metric) | 170 mm |