Learn More
Molecular Probes™ Fura-2, Pentasodium Salt, cell impermeant
Product Code. 10013212
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 10013212
Brand: Molecular Probes™ F6799
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Fura-2, Pentasodium Salt, cell impermeant
Fura-2, pentasodium salt is an intracellular calcium indicator that is ratiometric and UV light—excitable. This water-soluble salt form is useful for intracellular loading by microinjection, infusion from patch pipette or uptake induced by our Influx pinocytic cell-loading reagent (I-14402).
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope, Microphotometer, Flow Cytometer
Product Type Calcium Indicator
Dye Type Fluorescent Dye-Based
Quantity 1 mg
Shipping Condition Room Temperature