missing translation for 'onlineSavingsMsg'
Learn More

Furan-2-thiocarboxamide, 97%, Thermo Scientific™

Catalog Number p-7034168
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
17572-09-7
C5H5NOS
127.161
MFCD01935200
OUZNHXMJEBIDSZ-UHFFFAOYSA-N
furan-2-thiocarboxamide, 2-furancarbothioamide, amino-2-furylmethane-1-thione, furan-2-thioamide, 2-carbamothioylfuran, 2-furan-thiocarboxamide, furan-2-carbothioic acid amide
2777138
furan-2-carbothioamide
C1=COC(=C1)C(=S)N
This item is not returnable. View return policy
17572-09-7
C5H5NOS
127.161
MFCD01935200
OUZNHXMJEBIDSZ-UHFFFAOYSA-N
furan-2-thiocarboxamide, 2-furancarbothioamide, amino-2-furylmethane-1-thione, furan-2-thioamide, 2-carbamothioylfuran, 2-furan-thiocarboxamide, furan-2-carbothioic acid amide
2777138
furan-2-carbothioamide
C1=COC(=C1)C(=S)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 127.16
Percent Purity 97%
Chemical Name or Material Furan-2-thiocarboxamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'