missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl acetate, 97%

Product Code. 11438146
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11438146

Brand: Thermo Scientific Alfa Aesar A15285.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-17-6 | C7H8O3 | 140.138 g/mol

Furfuryl acetate is a flavoring ingredient. It was used in the synthesis of 5-acetoxymethyl-2-vinylfuran and 5-hydroxymethyl-2-vinylfuran via Vilsmeier-Haack and Wittig reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furfuryl acetate is a flavoring ingredient. It was used in the synthesis of 5-acetoxymethyl-2-vinylfuran and 5-hydroxymethyl-2-vinylfuran via Vilsmeier-Haack and Wittig reactions.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-17-6
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00003251
InChI Key CKOYRRWBOKMNRG-UHFFFAOYSA-N
Synonym furfuryl acetate, 2-furanmethanol, acetate, 2-furanmethanol acetate, 2-acetoxymethylfuran, furfuryl alcohol, acetate, acetic acid furfurylester, acetic acid furfuryl ester, 2-furanmethyl acetate, 2-furfuryl acetate, 2-furylmethyl acetate
PubChem CID 12170
IUPAC Name furan-2-ylmethyl acetate
SMILES CC(=O)OCC1=CC=CO1

Specifications

Density 1.118
Boiling Point 175°C to 177°C
Flash Point 65°C (149°F)
Refractive Index 1.463
Quantity 25 g
Beilstein 116128
Solubility Information Not miscible in water.
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material Furfuryl acetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.