Learn More
Bacharach™ Fyrite™ CO2 or O2 Analyzer Refill Kits
Click to view available options
Includes:
3 Bottles of fluid (7.6% CO2)
Concentration Measurement Range:
0 to 7.6% CO
Unit Size:
Pack of 3
Description
- Include two bottles of fluid, top gasket screws, filtering material 10-884-6 consists of three bottles of fluid only
Specifications
Specifications
| Concentration Measurement Range | 0 to 7.6% CO |
| Type | CO2 Analyzer Refill Kit |
| Includes | 3 Bottles of fluid (7.6% CO2) |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands, forearms, andother exposed areas thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see section 4 on this label).
- P330-Rinse mouth.
- P363-Wash contaminated clothing before reuse.
- P390-Absorb spillage to prevent material damage.
- P405-Store locked up.
- P406-Store in corrosive resistant/ container with a resistant inner liner.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : potassium hydroxide,alcohols C7-C9, C.I. acid red 14, 2-naphthalenol