Learn More
R&D Systems™ FzM1.8
Description
FzM1.8 is a frizzled 4 allosteric agonist (pEC50 = 6.4). Potentiates the β-catenin pathway in the absence of any WNT ligand, by biasing FZD4 signaling toward the FZD4/PI3K axis. Preserves stemness and promotes proliferation of undifferentiated cells.
Spécification
Spécification
| Nom chimique ou matériau | 3-Hydroxy-5-[[(2-Naphthalenylamino)carbonyl]amino]benzoic acid |
| CAS | 2204290-85-5 |
| Quantité | 50 mg |
| Gamme de produits | Tocris |
| Formule moléculaire | C18H14N2O4 |
| Pureté | 98% |