Learn More
Corning™ G418 Sulfate, Powder
Packaged in plastic bottles
Brand: Corning™ 61-234-RG
Additional Details : CAS : 69-52-3 Weight : 0.00500kg
Specifications
2° to 8°C | |
Powder | |
Antibiotic | |
5 g |
Packaged in plastic bottles
Brand: Corning™ 61-234-RG
Additional Details : CAS : 69-52-3 Weight : 0.00500kg
2° to 8°C | |
Powder | |
Antibiotic | |
5 g |