missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Gallein

Description
Inhibitor of G protein βγ subunit-dependent signaling. Blocks PI 3-kinase and Rac1 activation in HL60 cells and chemotaxis in HL60 differentiated cells.
Specifications
Specifications
| CAS | 2103-64-2 |
| Quantity | 50 mg |
| Molecular Formula | C20H12O7 |
| Formula Weight | 364.31 |
| Purity | >98% |
| Molecular Weight (g/mol) | 364.309 |
| Chemical Name or Material | Gallein |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?