Learn More
Emma Safety Footwear Galus Safety Shoes
Insulated with fine quality sheepskin to keep feet warm on the coldest of conditions. Nitrile rubber outsole won't yield if the temperature rises up to 300 degrees on the outside.
Brand:  Emma Safety Footwear MM58086045
Additional Details : Weight : 2.22000kg
 View more versions of this product
Product Code. 15958761
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • S2-Safety shoe for wet working conditions
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Fur-lined and insulated for warmth
Specifications
Specifications
EN ISO 20345:2011, EN 12568:2010
Black
Fur
45
Zip
Galus
Leather
Steel