Learn More
Keystone Cleanroom Products Gamma-Irradiated Ultra Fine Point Sharpie
Product Code. 17726909 Shop All Keystone Cleanroom Products Products
Click to view available options
Color:
Black
Blue
Unit Size:
Pack of 500
Product Code. 17726909
Brand: Keystone Cleanroom Products S01UFMARKBLUE
Only null left
Add to basket
Add to basket
Ready-to-use gamma irradiated pens and markers eliminate the risks associated with surface sanitization procedures.
  • Gamma irradiated
  • Lot controlled
  • Ready to use
  • Packaged in easy to open Tyvek™ peel pouches
  • Minimize contamination risks
  • Simplifies conformance to SOP's
  • Eliminate the need for wiping down contaminated writing instruments
Specifications
Tip Style Ultrafine
Product Type Cleanroom Marker
Color Blue
Autoclavable Not Autoclavable
Sterility Sterile
Material Plastic
Disposable Single-use
Certifications/Compliance COC, Certificate of Irradiation