Learn More
Berkshire™ Gamma™ Wipe SuperSorb™
Shop All Berkshire Products
Click to view available options
Dimensions (L x W):
30 x 30 cm
Unit Size:
Pack of 160
Description
It is composed of 100% continuous filament polyester knit fabric. Each package is gamma irradiated sterile to a 10-6 Sterility Assurance Level. This combination of properties provides Gamma Wipe Supersorb™ with the high level of cleanliness, abrasion resistance and chemical compatibility required for cleaning critical sterile processing environments.
Specifications
Specifications
| Material | Polyester |
| Construction | Knitted |
| Edges | Ultrasonically Sealed |
| Dimensions (L x W) | 30 x 30 cm |
| Certifications/Compliance (Europe) | ISO Class 3 - 5, Cleanroom Class 1 - 100, EU Grade A/B/C/D |