Learn More
Garcin™ Bactimains™ Liquid Soap for Handwashing
Use Garcin™ Bactimains™ KLiquid Soap for Handwashing in the workplace. The colorless, odorless liquid soap provides bacteriostatic and fungistatic protection from microbial contamination.
Brand: Garcin LB923690
Additional Details : Weight : 21.50000kg
Description
- 5L can
- Fragrance and dye-free, pH 6,5
- Active principles methylisothiazolinone, vegetable fatty acid derivatives, glycerin
- Bacteriostatic and fungistatic with respect to microbial contamination
- Included with metering pump
Specifications
Anti-microbial Liquid Soap | |
Liquid | |
6.5 | |
5 L |
Colorless | |
Dispenser | |
4 Pk. |
Safety and Handling
- X4 BACTIMAINS SOAP LIQUID 5L
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P102-Keep out of reach of children.
- P273-Avoid release to the environment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : N,N-DIMETHYLDODECYLAMINE N-OXIDE, glycerol