Learn More
Garcin™ Garcin™ Bactinyl Ionodore Disinfectant
Use Garcin™ Bactinyl Ionodore Disinfectant for multipurpose surface disinfection and cleaning and destruction of bacteria, fungi, mycobacteria, viruses and spores.
Brand: Garcin™ LB920800
Additional Details : CAS : 68424-85-1 Weight : 20.00000kg Transport : UN number : 3267 Chem class : 8 Pack group : II
Description
- CE-0398 certification
- 4 per pack
- 5L can
Specifications
Disinfectant | |
Can | |
5 L |
CE-0398 certification | |
4/Pk. |
Safety and Handling
- bactinyl PE indoor
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol, didecyldimethylammonium chloride