Learn More
Integra Biosciences™ Gas Safety Tubing
Product Code. 10247271
Click to view available options
Diameter (Metric):
10 mm
Unit Size:
2m
Product Code. 10247271
Brand: Integra Biosciences™ 140055
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Product Type Tube
For Use With (Equipment) Gas Cartridge Adapters
Length (Metric) 10 mm
Diameter (Metric) 10 mm