Learn More
Lenz Laborglasintrumente Gas Washing Bottles Acc. to Drechsel with Filter Disc
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05510014
Additional Details : Weight : 0.10000kg
Description
- Acc. to DIN 12596, complete with head (with filter disc)
- Ground joint connection: NS 29/32
- Material: DURAN™ tubing (head), borosilicate glass 3.3 (bottle)
Specifications
1000 mL | |
GL 14 connections | |
NS 29/32 |
Borosilicate Glass (3.3), Duran Tubing | |
0 |