Learn More
BD GasPak™ Supplies
Click to view available options
Type:
CO2 Indicator Strip
Catalyst Replacement Charges
Dry Anaerobic Indicator Strip
Tube Holder
Quantity:
1 Each
10 Pk.
100 Pk.
50 Pk.
Unit Size:
Each
Pack of 100
Pack of 50
Description
GasPak™ EZ Gas Generating Systems are complete, integrated systems that rapidly produce atmospheres suitable to support the primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.
Accessories include:
- CO2 Indicator Strips
- Dry Anaerobic Indicator Strips
- Tube Holder
- Catalyst Replacement Charges
- Catalyst Reaction Chamber
Specifications
Specifications
| Type | Tube Holder |
| Content And Storage | 15°C to 30°C |
| Description | Clear, plastic tube holder |
| For Use With (Application) | Produce atmospheres suitable to support the primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria |
| For Use With (Equipment) | BD GasPak™ |
| Holds | 16.5 x 128 Tubes (13) |
| Quantity | 1 Each |