Learn More
Trajan Scientific and Medical™ Replacement Mini Union Ferrules
Product Code. 12746348
Change view
Click to view available options
For Use With (Equipment):
0.1 to 0.32 mm I. D Columns
0.45 to 0.53 mm ID Columns
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12746348 0.1 to 0.32 mm I. D Columns Pack of 10
12736348 0.45 to 0.53 mm ID Columns Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12746348 Supplier Trajan Scientific and Medical™ Supplier No. 072630
Only null left
Add to basket
Add to basket
Leak-free system connections for improved chromatography for non-MS systems
  • Easy to use
  • Forms a reliable seal
  • Reusable
  • Soft material, forms a soft grip to the capillary column
  • Low emissions
Specifications
Type Ferrule
For Use With (Equipment) 0.1 to 0.32 mm I. D Columns
Material Graphite
Quantity 10 Pack