Learn More
Trajan Scientific and Medical™ Replacement Mini Union Ferrules
Click to view available options
For Use With (Equipment):
0.1 to 0.32 mm I. D Columns
0.45 to 0.53 mm ID Columns
Unit Size:
Pack of 10
Description
- Easy to use
- Forms a reliable seal
- Reusable
- Soft material, forms a soft grip to the capillary column
- Low emissions
Specifications
Specifications
| Type | Ferrule |
| For Use With (Equipment) | 0.45 to 0.53 mm ID Columns |
| Material | Graphite |
| Quantity | 10 Pack |