missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ GC inlet liner split/splitless tapered, FocusLiner for Shimadzu 2010 3.4mm I.D., 5mm O.D., 95mm length SGE

The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds

Brand:  Trajan Scientific and Medical™ 09205801

Additional Details : Weight : 0.10000kg

Product Code. 12923991

  • 70.20€ / Each
Estimated Shipment: 03-07-2024
to see stock.
Add to basket

Description

Description

  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with Shimadzu instruments
Specifications

Specifications

GC Inlet Liner
95 mm
5 mm
1 Ea.
3.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions