Learn More
Schmidlin™ GC Plus Rack Zero Air Generator
Product Code. 17748919
Encompass_Preferred
Click to view available options
Flow Rate:
15,000 mL/min.
1800 mL/min.
500 mL/min.
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 17748919
Brand: Schmidlin™ DBRGC15000EU
Regulatory: This product is under eco-contribution tax.
This generator produces a continuous supply of zero grade air.
  • Increase efficiency and improve safety and performance
  • On-demand supply 24/7
  • Purity: <0.1 ppm THC
  • Pressure: 4.5 to 10 barg (145 psig)
  • Proprietary platinum catalyst technology
  • Easy to install with complete 'plug and play' functionality
  • Ideal for all GC detector applications
  • Stable baseline with increased sensitivity and repeatability
  • Meets and exceeds requirements for the most demanding analyzers
Specifications
Depth (Metric) 500 cm
Flow Rate 15,000 mL/min.
Height (Metric) 13.3 cm
Max. Ambient Operating Temperature 35°C
Outlet Pressure 3.5 to 9 bar
Width (English) 19 in.
Width (Metric) 48.5 cm
Connections Inlet: 1/4 in., Outlet: 1/8 in.
Electrical Requirements 110/120 V at 60 Hz, 220/240 V at 50 Hz
Nitrogen Purity <0.1 ppm
Wattage 550 W