Learn More
Cytiva plasmidPrep Midi Flow Kit
For the rapid extraction and purification of high-quality plasmid DNA from medium scale cultures for use in transfection, sequencing, and enzymatic amplification and modification
Brand: Cytiva 28-9042-67
Additional Details : CAS : 9048-46-8 Weight : 0.50000kg Transport : UN number : 2790 Chem class : 8 Pack group : III
Includes: Fast-flow plasmid 250 columns; Resuspension, lysis, neutralization, wash, elution, and TE buffers; Full protocol booklet with a detachable, quick reference protocol card
Description
The kit consists of a high-performance, anion-exchange purification medium that combines excellent flow characteristics with an exceptional capacity and specificity for plasmid DNA.
- High column capacity allows for a proportional yield for input sample volume of 25 to 50mL (for high copy number plasmid) and up to 500mL for low copy number plasmid
- Provides plasmid DNA with excellent purity and low endotoxin levels
- Increased ease of use due to minimal pipetting volume changes, color-coded kit components, and pre-equilibrated columns
Specifications
illustra plasmidPrep Midi Flow Kit is designed for the purification of high yields of transfection-grade plasmid DNA. The kit allows for the purification of large yields of plasmid DNA that are proportional to the input sample volume. | |
Fast-flow genomic 250 columns, lyophilized Proteinase K powder, Lysis solutions, load and Elution Buffers, NAP-25 desalting column, spin column adapters, full protocol booklet, detachable, quick reference protocol card | |
illustra plasmidPrep Midi Flow Kit | |
150 mL Culture |
Used For the rapid extraction and purification of high-quality plasmid DNA from medium scale cultures for use in transfection, sequencing, and enzymatic amplification and modification | |
25 Preps | |
1.8 | |
Approx.215 min |
Safety and Handling
- Warning
Hazard Category
- Flammable liquid Category 3