Learn More
Biochrom Calibration Fluid for NanoVue Plus Spectrophotometer
Product Code. 10300225
Click to view available options
Detector Type:
Diode
Unit Size:
1 set
Product Code. 10300225
Brand: Biochrom 80214009
Use Calibration Fluid for NanoVue Plus Spectrophotometer to drop-and-measure sample plates for accurate and reliable quantitation of nucleic acid and protein sample with this Uracil solution in Sodium Azide.
  • Calibration Fluid for use with NanoVue Plus Spectrophotometer
Specifications
Description Pathlength calibration fluid
Detector Type Diode
Display No Display
Light Source Xenon Flash
Wavelength Range 260 to 280 nm
For Use With (Application) UV-Visible Spectrophotometer for Quick and Accurate Quantitation of Nucleic Acids and Protein Samples
For Use With (Equipment) NanoVue Plus Spectrophotometer
Product Identifier
  • Pathlength Calibration Fluid
Signal Word
  • Warning