Learn More
Melford Geldanamycin
Descrizione
Geldanamycin is a naturally occurring antibiotic derived from the bacterium Streptomyces hygroscopicus.
Geldanamycin is often used as a research tool to study cellular processes that are dependent on Hsp90. By inhibiting Hsp90, geldanamycin affects the stability and function of various client proteins associated with cell signaling, growth, and survival.
Researchers use geldanamycin to investigate the role of Hsp90 in the context of cancer, as many oncoproteins require Hsp90 for proper folding and stability.
Specifica
Specifica
| Chemical Name or Material | Geldanamycin |
| CAS | 30562-34-6 |
| Quantity | 500 μg |
| Molecular Formula | C29H40N2O9 |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.