missing translation for 'onlineSavingsMsg'
Learn More

Geldanamycin, 98+%, Thermo Scientific™

Benzoquinoid antibiotic that inhibits pp60src tyrosine kinase

Brand:  Thermo Scientific Alfa Aesar J63397.MB

Quantity 25mg

 View more versions of this product

Product Code. 15405649

  • POA / 25mg
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Geldanamycin binds to and inhibits the cytosolic chaperone functions of heat shock protein 90 (HSP90). HSP90 maintains the stability and functional shape of many oncogenic signaling proteins. It interferes with conformational maturation of proteins and the cellular stress response. In addition, it is a potent inhibitor of the nuclear hormone receptor family.

Solubility
Soluble in dimethyl sulfoxide at 100 mg/mL.

Notes
Store in a cool dry place. Keep away from oxidizing agents. Protect from heat.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

30562-34-6
560.644
QTQAWLPCGQOSGP-KDFVLELPSA-N
133688416
CC1CC(C(C(C=C(C(C(C=CC=C(C(=O)NC2=CC(=O)C(=C(C1)C2=O)OC)C)OC)OC(=O)N)C)C)O)OC
C29H40N2O9
MFCD00274570
NSC 122750; U-29135
[(3S,5R,6S,7R,8Z,11R,14E)-6-hydroxy-5,11,21-trimethoxy-3,7,9,15-tetramethyl-16,20,22-trioxo-17-azabicyclo[16.3.1]docosa-1(21),8,12,14,18-pentaen-10-yl] carbamate

Specifications

30562-34-6
25mg
MFCD00274570
Soluble in dimethyl sulfoxide at 100mg/mL.
CC1CC(C(C(C=C(C(C(C=CC=C(C(=O)NC2=CC(=O)C(=C(C1)C2=O)OC)C)OC)OC(=O)N)C)C)O)OC
560.644
560.64
Geldanamycin
Yellow to Orange
C29H40N2O9
NSC 122750; U-29135
QTQAWLPCGQOSGP-KDFVLELPSA-N
[(3S,5R,6S,7R,8Z,11R,14E)-6-hydroxy-5,11,21-trimethoxy-3,7,9,15-tetramethyl-16,20,22-trioxo-17-azabicyclo[16.3.1]docosa-1(21),8,12,14,18-pentaen-10-yl] carbamate
133688416
≥99%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.