missing translation for 'onlineSavingsMsg'
Learn More

Analytik Jena US UVP GelStudio PLUS and UVP GelStudio PLUS Touch

Analytik Jena US UVP GelStudio PLUS Imaging Systems include a high-performance 5MP imaging camera with wide aperture lens. Built to image DNA gels, protein gels, blue/white gels, plants, colorimetrics, and colony plates.

Brand:  Analytik Jena 849-00552-2

Additional Details : Poids : 39.00000kg

 View more versions of this product

Product Code. 15654879

  • 14340.00€ / Each


Expédition estimée
30-09-2020
Add to basket

Description

Description

  • Available as either a stand-alone touch screen operated imager or external computer operated system for laboratores with particlar IT/computer requirements
  • Maximizes laboratory bench space
Advanced technical specifications and capabilities:
  • High resolution, light sensitivity with 5.0 megapixel camera
  • f/1.2 lens for quick exposure speeds in low light and long exposure applications
  • Has overhead white, red, green and blue excitation LEDS for multiplex applications 
  • Use the UVP Visi-White™ LED Transilluminator or the economical alternative the UVP Visi-White™ Converter Plates for Coomassie blue, Silver Stains, and other trans white-light applications
  • Use the UVP Visi-Blue™ LED Transilluminator or the economical alternative the UVP Visi-Blue™ Converter Plate for blue excitation applications, including GelGreen and SYBR Green (460 to 470nm)
  • VisionWorks™ Acquisition and Analysis Software full package used on external computer
  • Features quantitative analysis and report generation
Designed for laboratory efficiency and ease-of-use:
  • UVP GelStudio touch includes an integrated 15.6in., wide touch-screen computer
  • Allows clear visuals of faint bands and finer details of images
  • Multi-tasking is easier to perform on wide screen, requiring multiple windows and files
  • Large and visible icons on VisionWorks™ touch Software, increases accessibility for improved and efficient workflow
  • Integrated computer has 500GB of hardware capacity to store large amounts of images, application presets and data
  • 7 USB ports available for use of keyboard, mouse and other accessories.
  • Stylus is included for simple and precise control
  • Darkroom features Thin-Line Transilluminator in slide-out tray, with uniform and high-intensity UV lighting in a compact form
  • Wide maximum illuminated imaging area of 25 x 26cm
  • Allows imaging of multiple gels, of various sizes
  • Thin-Line Transilluminator is designed with long-life UV lamps, for minimum service requirements
  • Thin UV lamp technology is instant-on with no need for warm-up
  • UV intensity switch “low/high” is included for optimal radiation for documentation or for preparative work
Automation features improves workflow in laboratories:
  • Image capture and enhancement is automated with application based icons
  • Used for quick selection of various gel applications
  • Custom application icons can be created for particular research needs
  • Large range of application settings are saved for various researchers performing experiments on one imaging system
  • Software controlled five-position filter wheel in slide-out tray
  • Allows for simple changeover to various emission wavelengths
  • All systems include unlimited copies of full package VisionWorks™ Acquisition and Analysis Software, with extensive quantitation and analysis features
  • Dedicated application and technical service teams available to provide customized service and support
  • Internal hardware, including camera, lens and computer is field accessible for troubleshooting and repairs
  • Expert technical support and application specialists
  • Three-year manufacturer warranty included with this system
  • IQ/OQ (Installation and Qualification) Documents and on-site support can be purchased
  •  

Certifications

CSA and CE certified

Warranty

3 year warranty

Specifications

Specifications

Monochrome CMOS camera of latest generation with 2592 x 1944 pixels resolution, 12 bit data depth, 16 bit file format. Motorized zoom lens 8 - 48 mm
15.6 in. Color touch screen
5-position filter wheel
VisionWorks Acquistion and Analysis Software
GelStudio PLUS Imaging Systems
230V
None
Low profile transilluminator with 25 cm x 26 cm UV filter size/302 nm UV/50 &/100 % intensity switch, on drawer. No operating costs by long-life UV bulbs. Optional converter plates: UV-to-White light, UV-to-Blue light, UV 302 nm-to-UV 365 nm, LED white l
Videos
Documents