Learn More
Agilent BioTek Gen5 Data Collection and Analysis Software
Product Code. 16562342 Shop All Agilent BioTek Products
Click to view available options
Model:
Gen5 AL2 Data Analysis Software
Gen5 AL3 Data Analysis Software
Gen5 AL4 Data Analysis Software
Gen5 IVD Data Analysis Software
Unit Size:
Each
Product Code. 16562342
Brand: Agilent BioTek GEN5AL4
Gen5 Data Collection and Analysis Software has a contemporary interface which provides easy access to all major functions of the software including reading parameters, plate layout, data reduction, online help, wizards, tutorials and sample protocols.
  • Quick launch buttons on the welcome screen provide links to recently used files, pre-programmed protocols and other key functions such as Quick Read and System Test
  • StepWise tools allow limitless combinations of procedural steps and data reduction criteria for a wide variety of applications in a logical and clearly defined sequence that is easily viewed on the screen
  • Analyze data on a single plate or across multiple plates in an experiment
  • Report and PowerExport builders are fast and flexible output tools to produce publication-ready reports using data matrices, statistics tables, protocols and plate information
  • For compliance to 21 CFR Part 11, Gen5 Secure provides system, data and protocol protection
  • Multiple user definitions and permission levels make it easy to comply with laboratory policies
System Requirements
  • Windows XP or Windows 2000 (Professional Editions)
  • PentiumIII-Class PC (or compatible) processor (500MHz or higher)
  • 512 MB RAM or higher
  • 2 GB hard drive space or higher
  • XGA resolution at 1024 x 768 or higher
  • CD-ROM readable drive
  • Keyboard
  • Mouse
  • Microsoft Internet Explorer v 5.0 or higher (for online help)
  • Microsoft Excel 2000 or later (for PowerExport feature)
TRUSTED_SUSTAINABILITY
Specifications
Model Gen5 AL4 Data Analysis Software
For Use With (Equipment) Imaging Multi-Mode Readers
Type Data Collection and Analysis Software