missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ Gqi5-NFAT-bla CHO-K1 Cells

Product Code. 10593514
Click to view available options
Quantity:
2 x 106 Cells
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10593514

Brand: Thermo Scientific™ K1725

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

The GeneBLAzer™ Gqi5-NFAT-bla CHO-K1 cell line contains the promiscuous Gα protein, Gqi5, stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line.

The GeneBLAzer™ Gqi5-NFAT-bla CHO-K1 cell line contains the promiscuous Gα protein, Gqi5, stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. NFAT-bla CHO-K1 cells (Cat. No. K1534) contain a beta-lactamase reporter gene under control of an NFAT response element stably integrated into CHO-K1 cells. The Gqi5-NFAT-bla CHO-K1 Parental Cell line is designed for the construction of Gi/o coupled GPCR cell based assays.

TRUSTED_SUSTAINABILITY

Specifications

Cell State Dividing Cells
Content And Storage GeneBLAzerTM Gqi5-NFAT-bla CHO-K1 cells are shipped on dry ice.

Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Detection Method Fluorescence
For Use With (Application) Pharma and biopharma, drug discovery and development, target and lead identification and validation, G-protein coupled receptor biology, cell-based GPCR reporter assays
Gene Symbol Gqi5
Includes Promiscuous Gα± protein, Gqi5, stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line
Product Line GeneBLAzer
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Sample Type CHO-K1
Type CHO-K1 Cells
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.