Learn More
Alla France™ Nitric/Sulfuric Acid Hydrometer
Product Code. 11743183
Click to view available options
Length (Metric):
300 mm
Unit Size:
Each
Product Code. 11743183
Brand: Alla France™ 3000TB100/20qp
Only null left
Add to basket
Add to basket
Reliably measure the specific gravity of your nitric or sulfuric acid solution. Alla France™ Nitric/Sulfuric Acid Hydrometer is a versatile, commercial-grade hydrometer.
  • Commercial grade hydrometer
  • Measures specific gravity of nitric or sulfuric acid
  • Range 1.000-2.000g/mL
Specifications
Quantity 1Each
Type General Purpose Acid Hydrometer
Accuracy ±0.010g/mL
Length (Metric) 300 mm
Specific Gravity Range 1.000 to 2.000 g/mL