missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Gentamicin Sulfate
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
25g
5g
Description
Gentamicin Sulfate is an aminoglycoside antibiotic that inhibits bacterial protein synthesis by binding to the bacterial ribosome.
Gentamicin Sulfate is an aminoglycoside antibiotic that inhibits bacterial protein synthesis by binding to the bacterial ribosome. This interference disrupts the translation process, leading to the production of faulty proteins and ultimately causing bacterial cell death.
It is frequently used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its inclusion in culture medium allows for the specific growth of bacteria lacking resistance to gentamicin.
Specifications
Specifications
| Chemical Name or Material | Gentamicin Sulfate |
| CAS | 1405-41-0 |
| Quantity | 10 g |
| Molecular Formula | C21H43N5O7-H2SO4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction