Learn More
Fisherbrand™ PTFE Giant Stir Bar
Shop All Fisherbrand Products
missing translation for 'orderingAttributeHoverText'
Length (Metric):
57 mm
80 mm
108 mm
110 mm
136 mm
159 mm
Diameter (Metric):
14 mm
24 mm
27 mm
36 mm
Shape:
Cylinder
Cylinder, Pivot ring
Triangle
missing translation for 'unitSize'
Each
Pack of 5
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
| Color | White |
| Description | PTFE encapsulated Alnico V magnet core |
| For Use With (Equipment) | Containers & Vessels / Stirrer Hot Plates |
| Diameter (Metric) | 14 mm |
| Length (Metric) | 80 mm |
| Quantity | 2/Pk. |
| Shape | Triangle |