Learn More
Gibco™ BGJb Medium
GREENER_CHOICE
Product Code. 11310892
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 11310892
Brand: Gibco™ 12591038
Requires
Supplementation
BGJb medium was developed for the growth of fetal rat long bones in a chemically defined medium.
BGJb medium was developed for the growth of fetal rat long bones in a chemically defined medium. BGJb is a medium modified by Sylvia Fitton-Jackson from the original BGJ medium developed by Biggers, Gwatkin, and Judah. BGJb medium creates favorable conditions for growth and calcification of fetal bones.
This BGJb medium is manufactured as follows:
With: L-glutamine, Phenol Red
Without: HEPES
The complete formulation is available.
Gibco™ BGJb medium contains higher levels of amino acids and glucose, and an increased buffering capacity to improve the conditions for growth and calcification of fetal bone.
Using BGJb medium
BGJb contains no proteins, lipids, or growth factors. Therefore, BGJb requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). BGJb uses a sodium bicarbonate buffer system (3.5 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Compliance
cGMP Manufacturing. Gibco BGJb medium is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
TRUSTED_SUSTAINABILITY
Specifications
Cell Type Fetal Bones
Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type BGJb Medium
Sterility Sterile-filtered
With Additives High Glucose, Glutamine, Phenol Red
Without Additives No HEPES, No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less
For Research Use Only. Not for use in diagnostic procedures. Not for human or animal therapeutic use. Uses other than the labeled intended use may be a violation of local law. For in vitro diagnostic use.